Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorobenzoyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2,4-Dichlorobenzoyl chloride
CAS 89-75-8
Molecular Formula C7H3Cl3O
Formula Weight 209.46
MDL Number MFCD00000661
UN Number UN3265
Beilstein 608234
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2081518

  SDS
alfa aesar™
B2081518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2081530

  SDS
alfa aesar™
B2081530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 209.46
Chemical Name or Material 2,4-Dichlorobenzoyl chloride
TSCA TSCA
Assay 98%
Molecular Formula C7H3Cl3O
MDL Number MFCD00000661
CAS 89-75-8
UN Number UN3265
Boiling Point 150°C (34mmHg)
Beilstein 608234
Melting Point 16° to 20°C
Flash Point 137°C (278°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
RTECS Number DM6636733
Quantity 50g

Applications
2,4-Dichlorobenzoyl chloride was used in the synthesis of biaryls by following the Suzuki coupling1. It was used for acylation in the synthesis of potent non-sarcosine-derived Gly T1 inhibitors. 2,4-Dichlorobenzoyl chloride was used in the synthesis of thioesters of 4-chlorobenzoate and 2,4-dichlorobenzoate.

Solubility
Reacts with water. Soluble in toluene.

Notes
Moisture sensitive. Store at room temperature. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.