Please login to your online account to display your discounted pricing

2,4-Dichlorobenzotrifluoride, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  166551000

 View more versions of this product

Catalog No. AC166551000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 215
Quantity 100g
Color Clear colorless
EINECS Number 206-278-1
TSCA TSCA
Density 1.4840g/mL
RTECS Number CZ5566877
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4805 to 1.4825 (20°C, 589nm)
Name Note 99%
Synonym 2, 4-DCBTF
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 4-Dichlorobenzotrifluoride
Assay Percent Range 99%
Molecular Formula C7H3Cl2F3
Linear Formula Cl2C6H3CF3
MDL Number MFCD00000582
CAS 320-60-5

Specifications

Chemical Name or Material 2, 4-Dichlorobenzotrifluoride
Name Note 99%
CAS 320-60-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H3Cl2F3
Linear Formula Cl2C6H3CF3
Formula Weight 215
MDL Number MFCD00000582
Synonym 2, 4-DCBTF
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents