missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorobenzotrifluoride, 99%, ACROS Organics™

Manufacturer:  Acros Organics 166550250

 View more versions of this product

Catalog No. AC166550250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4-Dichlorobenzotrifluoride
25g
320-60-5
100.0
Authentic
1.4805 to 1.4825 (20°C, 589nm)
99%
Cl2C6H3CF3
2,4-Dichloro-α,α,α-trifluorotoluene, 2, 4-Dichloro-α, α, 2, 4-Dichloro-α, α, 2, 4-DCBTF
C1=CC(=C(C=C1Cl)Cl)C(F)(F)F
214.996
215
Colorless
99%
98.5
1.4840g/mL
Glass Bottle
≥98.5%
C7H3Cl2F3
MFCD00000582
KALSHRGEFLVFHE-UHFFFAOYSA-N
2,4-dichloro-1-(trifluoromethyl)benzene
9443
Liquid

Chemical Identifiers

320-60-5
214.996
KALSHRGEFLVFHE-UHFFFAOYSA-N
9443
C7H3Cl2F3
MFCD00000582
2,4-Dichloro-α,α,α-trifluorotoluene, 2, 4-Dichloro-α, α, 2, 4-Dichloro-α, α, 2, 4-DCBTF
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-278-1

RTECSNumber : CZ5566877

TSCA : TSCA

SDS Documents