missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorobenzonitrile, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1011306

 View more versions of this product

Catalog No. AAA1011306



Description

Description

Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

Solubility
Soluble in Methanol. Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications

Specifications

2,4-Dichlorobenzonitrile
57°C to 61°C
MFCD00016373
UN3439
benzonitrile, 2,4-dichloro, 2,4-dichloro-benzonitrile, 2,4-dichlorobenzenecarbonitrile, acmc-209nrx, 2,4-dichloro benzonitrile, ksc352s3f
GRUHREVRSOOQJG-UHFFFAOYSA-N
2,4-dichlorobenzonitrile
81050
98%
6574-98-7
C7H3Cl2N
5 g
637738
Soluble in Methanol. Slightly soluble in water.
C1=CC(=C(C=C1Cl)Cl)C#N
172.008
172.01

Chemical Identifiers

6574-98-7
172.008
GRUHREVRSOOQJG-UHFFFAOYSA-N
81050
C1=CC(=C(C=C1Cl)Cl)C#N
C7H3Cl2N
MFCD00016373
benzonitrile, 2,4-dichloro, 2,4-dichloro-benzonitrile, 2,4-dichlorobenzenecarbonitrile, acmc-209nrx, 2,4-dichloro benzonitrile, ksc352s3f
2,4-dichlorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 229-493-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only