missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorobenzonitrile, 98%

Manufacturer:  Alfa Aesar™ A1011322

 View more versions of this product

Catalog No. AAA1011322



Description

Description

Applications
Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

Solubility
Soluble in Methanol. Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications

Specifications

57°C to 61°C
2,4-Dichlorobenzonitrile
C7H3Cl2N
UN3439
24-dichlorobenzenecarbonitrile, benzonitrile 24-dichloro, 24-dichloro-benzonitrile, 24-dichloro benzonitrile, acmc-209nrx, ksc352s3f
C1=CC(=C(C=C1Cl)Cl)C#N
172.008
172.01
100g
6574-98-7
MFCD00016373
637738
GRUHREVRSOOQJG-UHFFFAOYSA-N
2,4-dichlorobenzonitrile
81050
98%

Chemical Identifiers

6574-98-7
172.008
GRUHREVRSOOQJG-UHFFFAOYSA-N
81050
C1=CC(=C(C=C1Cl)Cl)C#N
C7H3Cl2N
MFCD00016373
24-dichlorobenzenecarbonitrile, benzonitrile 24-dichloro, 24-dichloro-benzonitrile, 24-dichloro benzonitrile, acmc-209nrx, ksc352s3f
2,4-dichlorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents