missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorobenzoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 160901000

 View more versions of this product

Catalog No. AC160901000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4-Dichlorobenzoic acid
Plastic bottle
>200.0°C
160.0°C to 164.0°C
98%
97.5
97.5% min. (HPLC)
Cl2C6H3CO2H
09, 342
Solubility in water: 0.36g/L (15°C). Other solubilities: soluble in alcohol,ether and acetone,insoluble in heptane
C1=CC(=C(C=C1Cl)Cl)C(=O)O
191.01
CHEBI:30748
Powder
Authentic
0.5% max. (K.F.)
White
100g
50-84-0
100.0
C7H4Cl2O2
MFCD00002414
2,4 dichlorobenzoic acid
ATCRIUVQKHMXSH-UHFFFAOYSA-N
2,4-dichlorobenzoic acid
5787
191.01
98%

Chemical Identifiers

50-84-0
191.01
ATCRIUVQKHMXSH-UHFFFAOYSA-N
5787
2,4-dichlorobenzoic acid
C7H4Cl2O2
MFCD00002414
2,4 dichlorobenzoic acid
CHEBI:30748
C1=CC(=C(C=C1Cl)Cl)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 200-067-8

RTECSNumber : DG6650000

TSCA : TSCA

SDS Documents