missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorobenzoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 160902500

Catalog No. AC160902500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 4-Dichlorobenzoic acid
50-84-0
100.0
Authentic
Plastic bottle
Cl2C6H3CO2H
250g
benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
ATCRIUVQKHMXSH-UHFFFAOYSA-N
2,4-dichlorobenzoic acid
5787
191.01
98%
97.5
>200.0°C
97.5% min. (HPLC)
C7H4Cl2O2
MFCD00002414
09, 342
Solubility in water: 0.36g/L (15°C). Other solubilities: soluble in alcohol,ether and acetone,insoluble in heptane
C1=CC(=C(C=C1Cl)Cl)C(=O)O
191.01
CHEBI:30748
98%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 200-067-8

RTECSNumber : DG6650000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only