Please login to your online account to display your discounted pricing

2,4-Dichlorobenzenesulfonyl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dichlorobenzenesulfonyl chloride
CAS 16271-33-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H3Cl3O2S
Linear Formula Cl(Cl)C6H3(SO2Cl)
Formula Weight 245.51
MDL Number MFCD00052712
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC356560010

  SDS
thermo fisher scientific
AC356560010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 245.51
Quantity 1g
Color White to light beige
Melting Point 54° to 58°C
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C6H3Cl3O2S
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
CAS 16271-33-3
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material 2, 4-Dichlorobenzenesulfonyl chloride
Linear Formula Cl(Cl)C6H3(SO2Cl)
MDL Number MFCD00052712
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%