Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorobenzenesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2,4-Dichlorobenzenesulfonyl chloride
CAS 16271-33-3
Molecular Formula C6H3Cl3O2S
Formula Weight 245.51
MDL Number MFCD00052712
UN Number UN3261
Beilstein 2806192
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1125914

 
alfa aesar™
A1125914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1125922

 
alfa aesar™
A1125922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.51
Chemical Name or Material 2,4-Dichlorobenzenesulfonyl chloride
Assay 98%
Molecular Formula C6H3Cl3O2S
MDL Number MFCD00052712
CAS 16271-33-3
UN Number UN3261
Beilstein 2806192
Melting Point 50° to 58°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number DB8927500
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.