Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichlorobenzamidoxime, 97%

$127.18 - $439.97

Chemical Identifiers

Chemical Name or Material 2,4-Dichlorobenzamidoxime
CAS 22179-80-2
Molecular Formula C7H6Cl2N2O
Formula Weight 205.04
MDL Number MFCD00178762
Synonym 2,4-Dichloro-N-hydroxybenzenecarboximidamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5067403

  SDS
alfa aesar™
H5067403
1g Each for $439.97

AAH50674MD

  SDS
alfa aesar™
H50674MD
250mg Each for $127.18
Description & Specifications

Specifications

Formula Weight 205.04
Chemical Name or Material 2,4-Dichlorobenzamidoxime
Assay 97%
Molecular Formula C7H6Cl2N2O
MDL Number MFCD00178762
CAS 22179-80-2
Synonym 2,4-Dichloro-N-hydroxybenzenecarboximidamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.