missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorobenzamide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 205590050

 View more versions of this product

Catalog No. AC205590050


 

Specifications Description & Specifications

Specifications

2, 4-Dichlorobenzamide
191° to 194°C
98%
97.5
Authentic
Crystalline Powder
98%
Cl2C6H3CONH2
MFCD00007974
White to off-white
5g
2447-79-2
100.0
Glass bottle
≥97.5%
C7H5Cl2NO
190.03
09, III, 1376

Chemical Identifiers

2447-79-2
100.0
C7H5Cl2NO
190.03
2, 4-Dichlorobenzamide, 98%
97.5
98%
Cl2C6H3CONH2
MFCD00007974
≥97.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 219-505-4

TSCA : TSCA