Please login to your online account to display your discounted pricing

2,4-Dichlorobenzamide, 98%, ACROS Organics™

$21.15 - $21.15

Chemical Identifiers

Chemical Name or Material 2, 4-Dichlorobenzamide
Name Note 98%
CAS 2447-79-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5Cl2NO
Linear Formula Cl2C6H3CONH2
Formula Weight 190.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205590050

  SDS
thermo fisher scientific
205590050
Glass bottle 5g Each for $21.15

AC205590250

  SDS
thermo fisher scientific
205590250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 190.03
Quantity 5g
Color White to off-white
EINECS Number 219-505-4
TSCA TSCA
Melting Point 191° to 194°C
Beilstein 09, III, 1376
CAS Max % 100.0
Chemical Name or Material 2, 4-Dichlorobenzamide
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H5Cl2NO
Linear Formula Cl2C6H3CONH2
MDL Number MFCD00007974
CAS 2447-79-2
CAS Min % 97.5