Please login to your online account to display your discounted pricing

2,4-Dichlorobenzaldehyde oxime, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB04865DA

 View more versions of this product

Catalog No. SPB04865DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.03
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Health Hazard 1 Warning
Chemical Name or Material 2, 4-Dichlorobenzaldehyde oxime
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 56843-28-8
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Assay Percent Range 97%
MDL Number MFCD00173694
Linear Formula Cl2C6H3CH=NOH
Molecular Formula C7H5Cl2NO

Specifications

Chemical Name or Material 2, 4-Dichlorobenzaldehyde oxime
CAS 56843-28-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5Cl2NO
Linear Formula Cl2C6H3CH=NOH
Formula Weight 190.03
MDL Number MFCD00173694
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents