missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorobenzaldehyde, 98%, ACROS Organics™

Manufacturer:  Acros Organics 164145000

 View more versions of this product

Catalog No. AC164145000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4-Dichlorobenzaldehyde
68°C to 73°C
98%
97.5
Authentic
≥97.5%
C7H4Cl2O
MFCD00003305
2, 4-DCAD
C1=CC(=C(C=C1Cl)Cl)C=O
175.008
175.01
Yellow through Beige
500g
874-42-0
100.0
Plastic bottle
98%
Cl2C6H3CHO
07, 236
YSFBEAASFUWWHU-UHFFFAOYSA-N
2,4-dichlorobenzaldehyde
13404
Crystalline Powder, Crystals and/or Chunks

Chemical Identifiers

874-42-0
175.008
YSFBEAASFUWWHU-UHFFFAOYSA-N
13404
C7H4Cl2O
MFCD00003305
2, 4-DCAD
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-861-1

TSCA : TSCA

SDS Documents