Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloroanisole, 99%

$27.57 - $342.04

Chemical Identifiers

Chemical Name or Material 2,4-Dichloroanisole
CAS 553-82-2
Molecular Formula C7H6Cl2O
Formula Weight 177.03
MDL Number MFCD00044772
Beilstein 1940358
Synonym 2,4-Dichloromethoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2004206

  SDS
alfa aesar™
B2004206
5g Each for $27.57

AAB2004222

  SDS
alfa aesar™
B2004222
100g Each for $342.04
Description & Specifications

Specifications

Formula Weight 177.03
Quantity 5g
Boiling Point 125°C (10mmHg)
Flash Point >110°C (230°F)
Melting Point 24° to 27°C
Beilstein 1940358
Chemical Name or Material 2,4-Dichloroanisole
Synonym 2,4-Dichloromethoxybenzene
MDL Number MFCD00044772
Molecular Formula C7H6Cl2O
CAS 553-82-2
TSCA TSCA
Assay 99%