missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichloroaniline, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1051830

 View more versions of this product

Catalog No. AAA1051830



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dichloroaniline
59°C to 63°C
242°C
C6H5Cl2N
250 g
386422
KQCMTOWTPBNWDB-UHFFFAOYSA-N
2,4-dichloroaniline
11123
162.02
554-00-7
1.567
149°C (300°F)
MFCD00007661
UN3442
2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german
C1=CC(=C(C=C1Cl)Cl)N
162.013
CHEBI:46635
99%

Chemical Identifiers

554-00-7
162.013
KQCMTOWTPBNWDB-UHFFFAOYSA-N
11123
2,4-dichloroaniline
C6H5Cl2N
MFCD00007661
2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german
CHEBI:46635
C1=CC(=C(C=C1Cl)Cl)N
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331-H373-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID

EINECSNumber : 209-057-8

RTECSNumber : BX2600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only