Please login to your online account to display your discounted pricing

2,4'-Dichloroacetophenone, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4'-Dichloroacetophenone
Name Note 99+%
CAS 937-20-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H6Cl2O
Linear Formula ClC6H4COCH2Cl
Formula Weight 189.04
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC136885000

  SDS
thermo fisher scientific
136885000
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.04
Assay ≥99.0%
Physical Form Crystalline Powder
Quantity 500g
Color White to off-white
EINECS Number 213-323-9
TSCA TSCA
Melting Point 99° to 102°C
CAS Max % 100.0
CAS Min % 99.0
Health Hazard 1 LACHRYMATOR
Synonym 2-Chloro-1-(4-chlorophenyl)ethanone
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99+%
Linear Formula ClC6H4COCH2Cl
CAS 937-20-2
Molecular Formula C8H6Cl2O
MDL Number MFCD00018926
Chemical Name or Material 2, 4'-Dichloroacetophenone
Name Note 99+%
Safety and Handling

LACHRYMATOR

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.