Please login to your online account to display your discounted pricing

2,4'-Dichloroacetophenone, 99+%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  136885000

 View more versions of this product

Catalog No. AC136885000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥99.0%
Formula Weight 189.04
Quantity 500g
Color White to off-white
EINECS Number 213-323-9
TSCA TSCA
Melting Point 99° to 102°C
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 2, 4'-Dichloroacetophenone
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C8H6Cl2O
Linear Formula ClC6H4COCH2Cl
MDL Number MFCD00018926
CAS 937-20-2
Synonym 2-Chloro-1-(4-chlorophenyl)ethanone
Health Hazard 1 LACHRYMATOR
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 4'-Dichloroacetophenone
Name Note 99+%
CAS 937-20-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H6Cl2O
Linear Formula ClC6H4COCH2Cl
Formula Weight 189.04
MDL Number MFCD00018926
Synonym 2-Chloro-1-(4-chlorophenyl)ethanone
Accessories

LACHRYMATOR

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents