Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Dichloroacetophenone, 98%

$61.64 - $548.46

Chemical Identifiers

Chemical Name or Material 2',4'-Dichloroacetophenone
CAS 2234-16-4
Molecular Formula C8H6Cl2O
Formula Weight 189.04
MDL Number MFCD00000581
Beilstein 637819
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1442814

  SDS
alfa aesar™
A1442814
25g Each for $61.64

AAA1442822

  SDS
alfa aesar™
A1442822
100g Each for $168.76

AAA1442836

  SDS
alfa aesar™
A1442836
500g Each for $548.46
Description & Specifications

Specifications

Formula Weight 189.04
Quantity 25g
Molecular Formula C8H6Cl2O
MDL Number MFCD00000581
CAS 2234-16-4
Health Hazard 1 Danger
Chemical Name or Material 2',4'-Dichloroacetophenone
Boiling Point 245° to 247°C
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Beilstein 637819
Flash Point >110°C (230°F)
Assay 98%
Melting Point 31° to 34°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.