missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4'-Dichloroacetophenone, 98+%

$213.92 - $213.92

Chemical Identifiers

CAS 937-20-2
Molecular Formula C8H6Cl2O
Formula Weight 189.04
MDL Number MFCD00018926
Beilstein 637861
Synonym 4-Chlorophenacyl chloride; alpha,4-Dichloroacetophenone
Chemical Name or Material 2,4'-Dichloroacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1595422 SDS Alfa Aesar™
A1595422
100g Each for $213.92
AAA1595414 SDS Alfa Aesar™
A1595414
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

937-20-2
189.04
637861
2,4'-Dichloroacetophenone
C8H6Cl2O
MFCD00018926
4-Chlorophenacyl chloride; alpha,4-Dichloroacetophenone

Specifications

100g
C8H6Cl2O
MFCD00018926
637861
4-Chlorophenacyl chloride; alpha,4-Dichloroacetophenone
≥98%
937-20-2
189.04
270°C
99° to 103°C
2,4'-Dichloroacetophenone

Safety and Handling

GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger