Please login to your online account to display your discounted pricing

2',4'-Dichloroacetophenone, 94%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2', 4'-Dichloroacetophenone
Name Note 94%
CAS 2234-16-4
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 94%
Molecular Formula C8H6Cl2O
Linear Formula Cl2C6H3COCH3
Formula Weight 189.04
Synonym 1-(2, 4-Dichlorophenyl)ethanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC176382500

  SDS
thermo fisher scientific
176382500
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.04
Quantity 250g
EINECS Number 218-780-8
TSCA TSCA
Density 1.32g/mL
Beilstein 07, II, 219
CAS Min % 92.0
CAS Max % 100.0
Chemical Name or Material 2', 4'-Dichloroacetophenone
Name Note 94%
Assay Percent Range 94%
Molecular Formula C8H6Cl2O
Linear Formula Cl2C6H3COCH3
CAS 2234-16-4
Synonym 1-(2, 4-Dichlorophenyl)ethanone
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.