Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Dichloroacetanilide, 98%

Chemical Identifiers

Chemical Name or Material 2',4'-Dichloroacetanilide
CAS 6975-29-7
Molecular Formula C8H7Cl2NO
Formula Weight 204.06g/mol
MDL Number MFCD00045181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2179509

 
alfa aesar™
B21795-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 146° to 148°C
Quantity 10g
Chemical Name or Material 2',4'-Dichloroacetanilide
CAS 6975-29-7
Assay 98%
Molecular Formula C8H7Cl2NO
Formula Weight 204.06g/mol
MDL Number MFCD00045181
RTECS Number AE1945000
TSCA TSCA