Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Dichloro-N-methylacetanilide, 97%

$356.64 - $356.64

Chemical Identifiers

Chemical Name or Material 2',4'-Dichloro-N-methylacetanilide
CAS 93646-27-6
Molecular Formula C9H9Cl2NO
Formula Weight 218.08g/mol
MDL Number MFCD01313704
Synonym N-(2,4-Dichlorophenyl)-N-methylacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2498314

 
alfa aesar™
B24983-14
25g Each for $356.64
Description & Specifications

Specifications

Melting Point 85° to 88°C
Quantity 25g
Chemical Name or Material 2',4'-Dichloro-N-methylacetanilide
CAS 93646-27-6
Assay 97%
Molecular Formula C9H9Cl2NO
Formula Weight 218.08g/mol
MDL Number MFCD01313704
Synonym N-(2,4-Dichlorophenyl)-N-methylacetamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.