Please login to your online account to display your discounted pricing

2,4-Dichloro-N-hydroxybenzenecarboximidoyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 4-Dichloro-N-hydroxybenzenecarboximidoyl chloride
CAS 29203-60-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4Cl3NO
Linear Formula Cl2C6H3CCl=NOH
Formula Weight 224.47
MDL Number MFCD00192643
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB06594EA

  SDS
fisher scientific company
SPB06594EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.47
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2, 4-Dichloro-N-hydroxybenzenecarboximidoyl chloride
Molecular Formula C7H4Cl3NO
MDL Number MFCD00192643
CAS 29203-60-9
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula Cl2C6H3CCl=NOH
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.