Please login to your online account to display your discounted pricing

2,4-Dichloro-α-methylbenzyl alcohol, 99+%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  188970500

 View more versions of this product

Catalog No. AC188970500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 191.06
Quantity 50g
Color Clear colorless
TSCA TSCA
EINECS Number 216-019-4
Density 1.29g/mL
Beilstein 06, II, 446
CAS Min % 99.0
CAS Max % 100.0
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99+%
Molecular Formula C8H8Cl2O
Assay Percent Range 99+%
Chemical Name or Material 2, 4-Dichloro-α-methylbenzyl alcohol
MDL Number MFCD00004511
CAS 1475-13-4
Linear Formula Cl2C6H3CH(CH3)OH

Specifications

Chemical Name or Material 2, 4-Dichloro-α-methylbenzyl alcohol
Name Note 99+%
CAS 1475-13-4
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H8Cl2O
Linear Formula Cl2C6H3CH(CH3)OH
Formula Weight 191.06
MDL Number MFCD00004511
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents