Please login to your online account to display your discounted pricing

2,4-Dichloro-α-methylbenzyl alcohol, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dichloro-α-methylbenzyl alcohol
Name Note 99+%
CAS 1475-13-4
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H8Cl2O
Linear Formula Cl2C6H3CH(CH3)OH
Formula Weight 191.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC188970100

  SDS
fisher scientific acros
188970100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC188970500

  SDS
thermo fisher scientific
188970500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 191.06
Physical Form Liquid
Quantity 10g
Color Clear colorless
Beilstein 06, II, 446
TSCA TSCA
Density 1.29g/mL
EINECS Number 216-019-4
CAS Max % 100.0
CAS Min % 99.0
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Linear Formula Cl2C6H3CH(CH3)OH
MDL Number MFCD00004511
Name Note 99+%
CAS 1475-13-4
Chemical Name or Material 2, 4-Dichloro-α-methylbenzyl alcohol
Assay Percent Range 99+%
Molecular Formula C8H8Cl2O
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.