missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-D-phenylalanine, 98%

$106.22 - $269.27

Chemical Identifiers

CAS 114872-98-9
Molecular Formula C9H9Cl2NO2
Formula Weight 234.09
MDL Number MFCD01860873
Synonym H-D-Phe(2,4-DiCl)-OH
Chemical Name or Material 2,4-Dichloro-D-phenylalanine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2726903 SDS Alfa Aesar™
H2726903
1g Each for $269.27
AAH27269MD SDS Alfa Aesar™
H27269MD
250mg Each for $106.22
Specifications Description & Specifications

Chemical Identifiers

114872-98-9
234.09
H-D-Phe(2,4-DiCl)-OH
C9H9Cl2NO2
MFCD01860873
2,4-Dichloro-D-phenylalanine

Specifications

114872-98-9
234.09
H-D-Phe(2,4-DiCl)-OH
−31° (c=0.5 in 0.5M NaOH)
98%
C9H9Cl2NO2
MFCD01860873
1g
2,4-Dichloro-D-phenylalanine