Please login to your online account to display your discounted pricing

2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 4-Dichloro-7H-pyrrolo[2, 3-d]pyrimidine
Assay Percent Range 95%
Molecular Formula C6H3Cl2N3
Formula Weight 188.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC454010010

  SDS
thermo fisher scientific
AC454010010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC454012500

  SDS
thermo fisher scientific
AC454012500
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.02
Quantity 1g
Assay Percent Range 95%
CAS 90213-66-4
Molecular Formula C6H3Cl2N3
Chemical Name or Material 2, 4-Dichloro-7H-pyrrolo[2, 3-d]pyrimidine