Please login to your online account to display your discounted pricing

2,4-Dichloro-6-(trifluoromethyl)pyrimidine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 4-Dichloro-6-(trifluoromethyl)pyrimidine
Assay Percent Range 95%
Molecular Formula C5HCl2F3N2
Formula Weight 216.98
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC453792500

  SDS
thermo fisher scientific
AC453792500
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC453790010

  SDS
thermo fisher scientific
AC453790010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2, 4-Dichloro-6-(trifluoromethyl)pyrimidine
Quantity 250mg
CAS 16097-64-6
Assay Percent Range 95%
Molecular Formula C5HCl2F3N2
Formula Weight 216.98