Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-6-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-6-(trifluoromethyl)aniline
CAS 62593-17-3
Molecular Formula C7H4Cl2F3N
Formula Weight 230.02
MDL Number MFCD00174313
UN Number UN3077
Synonym 2-Amino-3,5-dichlorobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2059003

  SDS
alfa aesar™
B2059003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2059014

  SDS
alfa aesar™
B2059014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2059006

  SDS
alfa aesar™
B2059006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.02
Boiling Point 49° to 52°C (0.4mmHg)
Health Hazard 3 GHS P Statement
P280h-P273-P305+P351+P338-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
CAS 62593-17-3
Assay 97%
Flash Point >110°C (230°F)
Synonym 2-Amino-3,5-dichlorobenzotrifluoride
MDL Number MFCD00174313
Chemical Name or Material 2,4-Dichloro-6-(trifluoromethyl)aniline
Molecular Formula C7H4Cl2F3N
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
UN Number UN3077
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P273-P305+P351+P338-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.