missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-6-methylquinazoline, 97%

Manufacturer:  Alfa Aesar™ H6301806

 View more versions of this product

Catalog No. AAH6301806



Specifications

Specifications

2,4-Dichloro-6-methylquinazoline
39576-82-4
quinazoline, 2,4-dichloro-6-methyl, 2,4-dichloro-6-methyl-quinazoline, 2,4 dichloro-6-methyl quinazoline, 6-methyl-2,4-dichloroquinazoline, pubchem23095
CC1=CC2=C(C=C1)N=C(N=C2Cl)Cl
213.061
213.06
97%
5g
C9H6Cl2N2
VZDVVLXYAOVNRW-UHFFFAOYSA-N
2,4-dichloro-6-methylquinazoline
14256483
Solid

Chemical Identifiers

39576-82-4
213.061
quinazoline, 2,4-dichloro-6-methyl, 2,4-dichloro-6-methyl-quinazoline, 2,4 dichloro-6-methyl quinazoline, 6-methyl-2,4-dichloroquinazoline, pubchem23095
2,4-dichloro-6-methylquinazoline
C9H6Cl2N2
VZDVVLXYAOVNRW-UHFFFAOYSA-N
14256483
CC1=CC2=C(C=C1)N=C(N=C2Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents