Please login to your online account to display your discounted pricing

2,4-Dichloro-6-methylpyrimidine, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  162821000

 View more versions of this product

Catalog No. AC162821000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 43° to 47°C
Color Yellow to yellow-orange
Quantity 100g
Chemical Name or Material 2, 4-Dichloro-6-methylpyrimidine
CAS 5424-21-5
CAS Min % 97.5
CAS Max % 100.0
Physical Form Yellow to yellow-orange crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C5H4Cl2N2
Formula Weight 163.01g/mol
MDL Number MFCD00006064
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact

Specifications

Chemical Name or Material 2, 4-Dichloro-6-methylpyrimidine
CAS 5424-21-5
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C5H4Cl2N2
Formula Weight 163.01g/mol
MDL Number MFCD00006064
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact

GHS P Statement Causes severe skin burns and eye damage.

SDS & Safety Documents