missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-6-methylpyridine, 97+%

$31.34 - $433.95

Chemical Identifiers

CAS 42779-56-6
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD08669834
InChI Key WUGTXQVNSRFDNV-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-methyl-pyridine
PubChem CID 11959083
IUPAC Name 2,4-dichloro-6-methylpyridine
SMILES CC1=NC(=CC(=C1)Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64248MD View Documents Alfa Aesar™
H64248MD
250mg Each for $31.34
 
AAH6424803 View Documents Alfa Aesar™
H6424803
1g Each for $108.81
 
AAH6424806 View Documents Alfa Aesar™
H6424806
5g Each for $433.95
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

42779-56-6
162.013
WUGTXQVNSRFDNV-UHFFFAOYSA-N
11959083
CC1=NC(=CC(=C1)Cl)Cl
C6H5Cl2N
MFCD08669834
2,4-dichloro-6-methyl-pyridine
2,4-dichloro-6-methylpyridine

Specifications

205°C to 206°C
42779-56-6
MFCD08669834
2,4-dichloro-6-methyl-pyridine
CC1=NC(=CC(=C1)Cl)Cl
162.013
162.01
2,4-Dichloro-6-methylpyridine
250mg
C6H5Cl2N
111084
WUGTXQVNSRFDNV-UHFFFAOYSA-N
2,4-dichloro-6-methylpyridine
11959083
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C