missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-6-methylpyridine, 97+%

Manufacturer:  Alfa Aesar™ H64248MD

 View more versions of this product

Catalog No. AAH64248MD



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

2,4-Dichloro-6-methylpyridine
250mg
C6H5Cl2N
111084
Slightly soluble in water.
CC1=NC(=CC(=C1)Cl)Cl
162.013
162.01
205°C to 206°C
42779-56-6
MFCD08669834
2,4-dichloro-6-methyl-pyridine, 2,4-dichloro-6-picoline, 2-methyl-4,6-dichloropyridine, d0ri6l, pubchem5465, pyridine,2,4-dichloro-6-methyl
WUGTXQVNSRFDNV-UHFFFAOYSA-N
2,4-dichloro-6-methylpyridine
11959083
≥97%

Chemical Identifiers

42779-56-6
162.013
WUGTXQVNSRFDNV-UHFFFAOYSA-N
11959083
CC1=NC(=CC(=C1)Cl)Cl
C6H5Cl2N
MFCD08669834
2,4-dichloro-6-methyl-pyridine, 2,4-dichloro-6-picoline, 2-methyl-4,6-dichloropyridine, d0ri6l, pubchem5465, pyridine,2,4-dichloro-6-methyl
2,4-dichloro-6-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents