Learn More
2,4-Dichloro-6-methylphenol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6431206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,4-Dichloro-6-methylphenol | |
52°C to 56°C | |
225°C (literature) | |
MFCD00019988 | |
UN3077 | |
4,6-dichloro-o-cresol, 4,6-dichloro-2-methylphenol, phenol, 2,4-dichloro-6-methyl, o-cresol, 4,6-dichloro, 4,6-dichlorocresol, phenol,2,4-dichloro-6-methyl, dichlorocresol, o-cresol,6-dichloro, 4,6-dichlor-o-kresol, acmc-1c0mk | |
CC1=CC(=CC(=C1O)Cl)Cl | |
177.024 | |
177.02 | |
Solid |
1570-65-6 | |
Brown | |
C7H6Cl2O | |
5 g | |
1864865 | |
WJQZZLQMLJPKQH-UHFFFAOYSA-N | |
2,4-dichloro-6-methylphenol | |
15292 | |
98% |
Chemical Identifiers
1570-65-6 | |
177.024 | |
WJQZZLQMLJPKQH-UHFFFAOYSA-N | |
2,4-dichloro-6-methylphenol |
C7H6Cl2O | |
MFCD00019988 | |
15292 | |
CC1=CC(=CC(=C1O)Cl)Cl |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 216-382-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only