Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-6,7-dimethoxyquinazoline, 97%

$96.82 - $309.39

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-6,7-dimethoxyquinazoline
CAS 27631-29-4
Molecular Formula C10H8Cl2N2O2
Formula Weight 259.09
MDL Number MFCD00051733
Beilstein 190331
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0605106

 
alfa aesar™
L0605106
5g Each for $96.82

AAL0605114

 
alfa aesar™
L0605114
25g Each for $309.39
Description & Specifications

Specifications

Formula Weight 259.09
Chemical Name or Material 2,4-Dichloro-6,7-dimethoxyquinazoline
Assay 97%
CAS 27631-29-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00051733
Beilstein 190331
Melting Point 175° to 179°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Molecular Formula C10H8Cl2N2O2

Applications
2,4-Dichloro-6,7-dimethoxyquinazoline is used as an intermediate in the preparation of potential inhibitors of epidermal growth factor receptor kinases. It is also used as an intermediate in the preparation of terazosin.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.