missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichloro-5-nitropyrimidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435290010

 View more versions of this product

Catalog No. AC435290010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 4-Dichloro-5-nitropyrimidine
Glass bottle
Yellow
1g
96.0
96% min. (GC)
MFCD00127867
INUSQTPGSHFGHM-UHFFFAOYSA-N
2,4-dichloro-5-nitropyrimidine
521266
Liquid or Low Melting Solid
Authentic
135°C (17.0 Torr)
29°C to 30°C
49845-33-2
100.0
C4HCl2N3O2
2, 4-dichloro-5-nitropyrimidine, 2,4-dichloro-5-nitro pyrimidine, 2,4-dichloro-5-nitro-pyrimidine, 2,4-dichloro5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, acmc-1ann4, ksc236i7h, pubchem7079, pyrimidine, 2,4-dichloro-5-nitro, zlchem 540
C1=C(C(=NC(=N1)Cl)Cl)[N+](=O)[O-]
193.98
193.98
97%

Chemical Identifiers

49845-33-2
193.98
INUSQTPGSHFGHM-UHFFFAOYSA-N
521266
C1=C(C(=NC(=N1)Cl)Cl)[N+](=O)[O-]
C4HCl2N3O2
MFCD00127867
2, 4-dichloro-5-nitropyrimidine, 2,4-dichloro-5-nitro pyrimidine, 2,4-dichloro-5-nitro-pyrimidine, 2,4-dichloro5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, acmc-1ann4, ksc236i7h, pubchem7079, pyrimidine, 2,4-dichloro-5-nitro, zlchem 540
2,4-dichloro-5-nitropyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents