Learn More
2,4-Dichloro-5-nitropyrimidine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 435290050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Dichloro-5-nitropyrimidine | |
96.0 | |
135°C (17.0 Torr) | |
96% min. (GC) | |
C4HCl2N3O2 | |
5g | |
INUSQTPGSHFGHM-UHFFFAOYSA-N | |
2,4-dichloro-5-nitropyrimidine | |
521266 | |
97% |
49845-33-2 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00127867 | |
2,4-dichloro-5-nitro-pyrimidine, pyrimidine, 2,4-dichloro-5-nitro, 2, 4-dichloro-5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, 2,4-dichloro-5-nitro pyrimidine, zlchem 540, pubchem7079, acmc-1ann4, ksc236i7h, 2,4-dichloro5-nitropyrimidine | |
[O-][N+](=O)C1=CN=C(Cl)N=C1Cl | |
193.97 | |
193.98 |
Chemical Identifiers
49845-33-2 | |
193.97 | |
INUSQTPGSHFGHM-UHFFFAOYSA-N | |
521266 | |
[O-][N+](=O)C1=CN=C(Cl)N=C1Cl |
C4HCl2N3O2 | |
MFCD00127867 | |
2,4-dichloro-5-nitro-pyrimidine, pyrimidine, 2,4-dichloro-5-nitro, 2, 4-dichloro-5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, 2,4-dichloro-5-nitro pyrimidine, zlchem 540, pubchem7079, acmc-1ann4, ksc236i7h, 2,4-dichloro5-nitropyrimidine | |
2,4-dichloro-5-nitropyrimidine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only