missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-nitrophenol, 98%

$96.84 - $388.61

Chemical Identifiers

CAS 39489-77-5
Molecular Formula C6H3Cl2NO3
Molecular Weight (g/mol) 207.994
MDL Number MFCD03093072
InChI Key OFAPWTOOMSVMIU-UHFFFAOYSA-N
Synonym phenol, 2,4-dichloro-5-nitro, 2,4-dichloro-5-nitrobenzenol, 2,4-dichloro-5-nitro-phenol, dichloronitrobenzenol, acmc-1afv8, ksc495i3b
PubChem CID 181635
IUPAC Name 2,4-dichloro-5-nitrophenol
SMILES C1=C(C(=CC(=C1O)Cl)Cl)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1836906 View Documents Alfa Aesar™
L1836906
5g Each for $96.84
 
AAL1836914 View Documents Alfa Aesar™
L1836914
25g Each for $388.61
 
Specifications

Chemical Identifiers

39489-77-5
207.994
OFAPWTOOMSVMIU-UHFFFAOYSA-N
181635
C1=C(C(=CC(=C1O)Cl)Cl)[N+](=O)[O-]
C6H3Cl2NO3
MFCD03093072
phenol, 2,4-dichloro-5-nitro, 2,4-dichloro-5-nitrobenzenol, 2,4-dichloro-5-nitro-phenol, dichloronitrobenzenol, acmc-1afv8, ksc495i3b
2,4-dichloro-5-nitrophenol

Specifications

97°C to 102°C
39489-77-5
MFCD03093072
1733418
OFAPWTOOMSVMIU-UHFFFAOYSA-N
2,4-dichloro-5-nitrophenol
181635
98%
5g
C6H3Cl2NO3
UN2811
phenol, 2,4-dichloro-5-nitro, 2,4-dichloro-5-nitrobenzenol, 2,4-dichloro-5-nitro-phenol, dichloronitrobenzenol, acmc-1afv8, ksc495i3b
C1=C(C(=CC(=C1O)Cl)Cl)[N+](=O)[O-]
207.994
208
2,4-Dichloro-5-nitrophenol
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.