Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-nitrobenzotrifluoride, 97%

$42.19 - $468.61

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-5-nitrobenzotrifluoride
CAS 400-70-4
Molecular Formula C7H2Cl2F3NO2
Formula Weight 260
MDL Number MFCD00007073
Beilstein 2467459
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1611106

  SDS
alfa aesar™
A1611106
5g Each for $42.19

AAA1611114

  SDS
alfa aesar™
A1611114
25g Each for $136.97

AAA1611122

  SDS
alfa aesar™
A1611122
100g Each for $468.61
Description & Specifications

Specifications

Formula Weight 260
Chemical Name or Material 2,4-Dichloro-5-nitrobenzotrifluoride
Beilstein 2467459
Quantity 5g
Molecular Formula C7H2Cl2F3NO2
Assay 97%
MDL Number MFCD00007073
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Melting Point 53° to 57°C
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 400-70-4
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.