missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichloro-5-methylpyrimidine, 98%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 98%
Molecular Formula C5H4Cl2N2
Formula Weight 163.01
Chemical Name or Material 2, 4-Dichloro-5-methylpyrimidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC398290010 SDS Thermo Fisher Scientific
AC398290010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398290050 SDS Thermo Fisher Scientific
AC398290050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

98%
C5H4Cl2N2
163.01
2, 4-Dichloro-5-methylpyrimidine

Specifications

163.01
1.3900g/mL
98%
Glass bottle
1g
C5H4Cl2N2
1780-31-0
2, 4-Dichloro-5-methylpyrimidine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger