missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-methylpyrimidine, 98%

Manufacturer:  Alfa Aesar™ H5520603

 View more versions of this product

Catalog No. AAH5520603



Description

Description

Applications
2,4-Dichloro-5-methylpyrimidine, is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine. It can react with piperidineto produce 2-chloro-5-methyl-4-piperidin-1-yl-pyrimidine. This reaction will need solvent dioxane.

Solubility
Soluble in chloroform, ether, ethyl acetate and toluene

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2,4-Dichloro-5-methylpyrimidine
113°C (235°F)
26°C to 28°C
1780-31-0
MFCD00023197
2, 4-dichloro-5-methylpyrimidine
DQXNTSXKIUZJJS-UHFFFAOYSA-N
2,4-dichloro-5-methylpyrimidine
74508
98%
1.39
108°C to 109°C (11mmHg)
1g
C5H4Cl2N2
UN3261
Soluble in chloroform,ether,ethyl acetate and toluene
CC1=CN=C(N=C1Cl)Cl
163.001
163.01

Chemical Identifiers

1780-31-0
163.001
DQXNTSXKIUZJJS-UHFFFAOYSA-N
74508
CC1=CN=C(N=C1Cl)Cl
C5H4Cl2N2
MFCD00023197
2, 4-dichloro-5-methylpyrimidine
2,4-dichloro-5-methylpyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 217-227-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents