missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-methylpyridine, 96%

Manufacturer:  Alfa Aesar™ H61634MD

 View more versions of this product

Catalog No. AAH61634MD



Description

Description

Applications
2,4-Dichloro-5-methylpyridine is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
Specifications

Specifications

2,4-Dichloro-5-methylpyridine
250mg
C6H5Cl2N
2,4-bis chloranyl-5-methyl-pyridine, 2,4-dichloro-5-methyl pyridine, 2,4-dichloro-5-methyl-pyridine, 2,4-dichloro-5-picoline, acmc-209lv3, ksc495i9n, pubchem5464, pyridine, 2,4-dichloro-5-methyl
DUPJNQNLAKJWKU-UHFFFAOYSA-N
2,4-dichloro-5-methylpyridine
12643718
Liquid
Colorless
56961-78-5
MFCD08704813
Slightly soluble in water.
CC1=CN=C(C=C1Cl)Cl
162.013
162.02
96%

Chemical Identifiers

56961-78-5
162.013
DUPJNQNLAKJWKU-UHFFFAOYSA-N
12643718
CC1=CN=C(C=C1Cl)Cl
C6H5Cl2N
MFCD08704813
2,4-bis chloranyl-5-methyl-pyridine, 2,4-dichloro-5-methyl pyridine, 2,4-dichloro-5-methyl-pyridine, 2,4-dichloro-5-picoline, acmc-209lv3, ksc495i9n, pubchem5464, pyridine, 2,4-dichloro-5-methyl
2,4-dichloro-5-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents