missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-methoxypyrimidine, 97%

Manufacturer:  Alfa Aesar™ H5615309

 View more versions of this product

Catalog No. AAH5615309



Description

Description

Applications
It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer’s disease.

Solubility
Slightly soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

2,4-Dichloro-5-methoxypyrimidine
10g
C5H4Cl2N2O
2,4 dichloro-5-methoxypyrimidine
ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
2,4-dichloro-5-methoxypyrimidine
601401
97%
66°C to 70°C
19646-07-2
MFCD02685670
Slightly soluble in water.
COC1=CN=C(N=C1Cl)Cl
179
179

Chemical Identifiers

19646-07-2
179
ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
601401
COC1=CN=C(N=C1Cl)Cl
C5H4Cl2N2O
MFCD02685670
2,4 dichloro-5-methoxypyrimidine
2,4-dichloro-5-methoxypyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents