Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-(iodomethyl)pyrimidine, 98%

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-5-(iodomethyl)pyrimidine
CAS 7627-44-3
Formula Weight 288.9
MDL Number MFCD12827832
Beilstein 512189
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH64805MD

  SDS
alfa aesar™
H64805MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 52° to 53°C
Formula Weight 288.9
Quantity 250mg
Assay 98%
Sensitivity Light Sensitive
CAS 7627-44-3
Chemical Name or Material 2,4-Dichloro-5-(iodomethyl)pyrimidine
Physical Form Solid
MDL Number MFCD12827832
Beilstein 512189