Please login to your online account to display your discounted pricing

2,4-Dichloro-5-fluoropyrimidine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dichloro-5-fluoropyrimidine
CAS 2927-71-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4HCl2FN2
Formula Weight 166.97
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC443220050

  SDS
thermo fisher scientific
AC443220050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC443220010

  SDS
thermo fisher scientific
AC443220010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder or Low Melting Solid
Assay ≥96.0%
Formula Weight 166.97
Quantity 5g
Color White to pale brown
UN Number 3261
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C4HCl2FN2
CAS 2927-71-1
Chemical Name or Material 2, 4-Dichloro-5-fluoropyrimidine
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.