Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-fluorophenylacetic acid, 97%

$62.84 - $155.20

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-5-fluorophenylacetic acid
CAS 10129-77-8
Molecular Formula C8H5Cl2FO2
Formula Weight 223.03
MDL Number MFCD01631383
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2659503

  SDS
alfa aesar™
H2659503
1g Each for $155.20

AAH26595MD

  SDS
alfa aesar™
H26595MD
250mg Each for $62.84
Description & Specifications

Specifications

Formula Weight 223.03
Molecular Formula C8H5Cl2FO2
Quantity 1g
Chemical Name or Material 2,4-Dichloro-5-fluorophenylacetic acid
MDL Number MFCD01631383
Assay 97%
CAS 10129-77-8
Melting Point 120° to 122°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.