Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-fluorobenzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-5-fluorobenzoyl chloride
CAS 86393-34-2
Molecular Formula C7H2Cl3FO
Formula Weight 227.45
MDL Number MFCD00075341
UN Number UN3265
Beilstein 4989077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2351403

  SDS
alfa aesar™
B2351403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2351406

  SDS
alfa aesar™
B2351406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.45
Chemical Name or Material 2,4-Dichloro-5-fluorobenzoyl chloride
Assay 97%
Molecular Formula C7H2Cl3FO
MDL Number MFCD00075341
CAS 86393-34-2
UN Number UN3265
Boiling Point 143° to 144°C (35mmHg)
Beilstein 4989077
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H317-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280-P262-P273-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P262-P273-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H317-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.