Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-5-fluorobenzamide, 97+%

$48.66 - $168.76

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-5-fluorobenzamide
CAS 135748-33-3
Molecular Formula C7H4Cl2FNO
Formula Weight 208.02
MDL Number MFCD01631478
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2440103

  SDS
alfa aesar™
B2440103
1g Each for $48.66

AAB2440106

  SDS
alfa aesar™
B2440106
5g Each for $168.76
Description & Specifications

Specifications

Formula Weight 208.02
Quantity 1g
Chemical Name or Material 2,4-Dichloro-5-fluorobenzamide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H4Cl2FNO
Health Hazard 1 Warning
MDL Number MFCD01631478
CAS 135748-33-3
Melting Point 158° to 160°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.