Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Dichloro-5'-fluoroacetophenone, 97%

$78.81 - $737.31

Chemical Identifiers

Chemical Name or Material 2',4'-Dichloro-5'-fluoroacetophenone
CAS 704-10-9
Molecular Formula C8H5Cl2FO
Formula Weight 207.03
MDL Number MFCD00077499
Beilstein 3090042
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2215306

  SDS
alfa aesar™
B2215306
5g Each for $78.81

AAB2215314

  SDS
alfa aesar™
B2215314
25g Each for $246.28

AAB2215322

  SDS
alfa aesar™
B2215322
100g Each for $737.31
Description & Specifications

Specifications

Formula Weight 207.03
Chemical Name or Material 2',4'-Dichloro-5'-fluoroacetophenone
Assay 97%
Molecular Formula C8H5Cl2FO
MDL Number MFCD00077499
CAS 704-10-9
Boiling Point 134° to 135°C (15mmHg)
Beilstein 3090042
Melting Point 34° to 36°C
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.